siRNA Details
| Gene Symbol: Cpne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153507.2 | NP_705727.1 | 8 | 905 | ||
| GenBank mRNA | AK160238.1 | - | 8 | 895 | ||
| BC022128.1 | - | 8 | 866 | 5097527 | ||
| BC023348.1 | - | 3 | 203 | 5012573 | ||
| BC031801.1 | - | 8 | 905 | 3985959 | ||

Cpne2
Genomic Map