siRNA Details
| Gene Symbol: Ccdc102a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033533.3 | NP_001028705.2 | 4 | 1135 | ||
| GenBank mRNA | AK144530.1 | - | 4 | 1088 | ||
| AK165668.1 | - | 4 | 1092 | |||
| BC107374.2 | - | 3 | 592 | |||
| BC107375.1 | - | 3 | 595 | |||
| BC114992.1 | - | 3 | 597 | |||
| BC139307.1 | - | 3 | 980 | 8862329 | ||
| BC139308.1 | - | 3 | 980 | 8862330 | ||

Ccdc102a
Genomic Map