siRNA Details
| Gene Symbol: Ces4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146213.2 | NP_666325.2 | 12 | 1558 | ||
| GenBank mRNA | AK037191.1 | - | 12 | 1481 | ||
| BC026374.1 | - | 12 | 1558 | 4163362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ces4a| Gene Symbol: Ces4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146213.2 | NP_666325.2 | 12 | 1558 | ||
| GenBank mRNA | AK037191.1 | - | 12 | 1481 | ||
| BC026374.1 | - | 12 | 1558 | 4163362 | ||