siRNA Details
| Gene Symbol: D230025D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145604.2 | NP_663579.2 | 7 | 748 | ||
| GenBank mRNA | AK156120.1 | - | 7 | 750 | ||
| AK166937.1 | - | 6 | 1935 | |||
| AK213296.1 | - | 5 | 307 | |||
| AK218692.1 | - | 6 | 391 | |||
| AK218947.1 | - | 3 | 195 | |||
| BC006874.1 | - | 7 | 631 | 2646515 | ||
| BC057919.1 | - | 7 | 737 | 5134302 | ||

D230025D16Rik
Genomic Map