siRNA Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172760.3 | NP_766348.3 | 5 | 435 | ||
| GenBank mRNA | AK037067.1 | - | 5 | 428 | ||
| AK157875.1 | - | 373 | ||||
| BC018516.1 | - | 3 | 311 | 3155913 | ||
| BC026617.1 | - | 1 | 292 | |||
| BC058752.1 | - | 5 | 418 | 6485393 | ||
| BC138887.1 | - | 5 | 392 | 8861909 | ||

Elmo3
Genomic Map