siRNA Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172760.3 | NP_766348.3 | 15, 16 | 1723 | ||
| GenBank mRNA | AK037067.1 | - | 15, 16 | 1716 | ||
| BC018516.1 | - | 13, 14 | 1612 | 3155913 | ||
| BC026617.1 | - | 10, 11 | 1668 | |||
| BC058752.1 | - | 15, 16 | 1706 | 6485393 | ||
| BC138887.1 | - | 15, 16 | 1680 | 8861909 | ||

Elmo3
Genomic Map