siRNA Details
| Gene Symbol: Elmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172760.3 | NP_766348.3 | 17 | 1916 | ||
| GenBank mRNA | AK037067.1 | - | 17 | 1909 | ||
| BC018516.1 | - | 15 | 1805 | 3155913 | ||
| BC026617.1 | - | 12 | 1861 | |||
| BC058752.1 | - | 17 | 1899 | 6485393 | ||
| BC138887.1 | - | 17 | 1873 | 8861909 | ||

Elmo3
Genomic Map