siRNA Details
| Gene Symbol: Lrrc29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177449.3 | NP_803232.2 | 3 | 1063 | ||
| GenBank mRNA | AK041606.1 | - | 3 | 1048 | ||
| AK042435.1 | - | 3 | 1048 | |||
| AK148120.1 | - | 3 | 1065 | |||
| AK170400.1 | - | 3 | 1063 | |||

Lrrc29
Genomic Map