siRNA Details
| Gene Symbol: Nrn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301174.1 | NP_001288103.1 | 2 | 151 | ||
| NM_175024.4 | NP_778189.1 | 2 | 154 | |||
| GenBank mRNA | AK080711.1 | - | 2 | 149 | ||
| AK090312.1 | - | 2 | 152 | |||
| BC100315.1 | - | 2 | 154 | 30931461 | ||
| CK030777.1 | - | 2 | 135 | |||
| DQ176852.1 | - | 2 | 141 | |||

Nrn1l
Genomic Map