siRNA Details
| Gene Symbol: Txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175646.3 | NP_783577.2 | 4 | 453 | ||
| GenBank mRNA | AK079533.1 | - | 4 | 552 | ||
| AK085226.1 | - | 4 | 434 | |||
| BC094379.1 | - | 4 | 469 | 30541506 | ||
| BC132373.1 | - | 3 | 378 | 40130650 | ||
| BC138333.1 | - | 3 | 378 | 8861353 | ||

Txnl4b
Genomic Map