siRNA Details
| Gene Symbol: Fuk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172283.3 | NP_758487.2 | 22 | 2956 | ||
| GenBank mRNA | AJ534942.1 | - | 22 | 2927 | ||
| AK049153.1 | - | 11 | 1785 | |||
| AK131165.1 | - | 11 | 4306 | |||
| AK161763.1 | - | 22 | 3060 | |||
| AK164340.1 | - | 22 | 3023 | |||
| AY223865.1 | - | 22 | 2927 | |||

Fuk
Genomic Map