siRNA Details
| Gene Symbol: Ddx19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190786.1 | NP_001177715.1 | 6 | 498 | ||
| NM_001190800.1 | NP_001177729.1 | 6 | 541 | |||
| NM_172284.5 | NP_758488.1 | 6 | 541 | |||
| GenBank mRNA | AK013362.1 | - | 6 | 536 | ||
| AK033235.1 | - | 6 | 499 | |||
| AK147499.1 | - | 6 | 536 | |||
| BC025594.1 | - | 6 | 501 | 5342859 | ||

Ddx19b
Genomic Map