siRNA Details
| Gene Symbol: Spg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153176.4 | NP_694816.3 | 2 | 273 | ||
| GenBank mRNA | AF512565.1 | - | 2 | 305 | ||
| AF547215.1 | - | 2 | 602 | |||
| AK145540.1 | - | 2 | 274 | |||
| BC051051.1 | - | 2 | 207 | |||
| BC055488.1 | - | 118 | ||||
| BC138141.1 | - | 2 | 500 | 8861160 | ||

Spg7
Genomic Map