siRNA Details
| Gene Symbol: Nup133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172288.2 | NP_758492.2 | 23 | 3250 | ||
| GenBank mRNA | AK029317.1 | - | 23 | 3234 | ||
| BC023915.1 | - | 24 | 3261 | 5323157 | ||
| BC026845.1 | - | 23 | 3233 | 4235408 | ||
| BC033476.1 | - | 23 | 3245 | |||
| BC058707.1 | - | 4 | 2039 | |||

Nup133
Genomic Map