siRNA Details
| Gene Symbol: BC021891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145608.2 | NP_663583.2 | 7 | 1866 | ||
| GenBank mRNA | AK220424.1 | - | 7 | 1866 | ||
| BC021891.1 | - | 7 | 1859 | 3156324 | ||
| BC046448.1 | - | 4 | 434 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC021891| Gene Symbol: BC021891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145608.2 | NP_663583.2 | 7 | 1866 | ||
| GenBank mRNA | AK220424.1 | - | 7 | 1866 | ||
| BC021891.1 | - | 7 | 1859 | 3156324 | ||
| BC046448.1 | - | 4 | 434 | |||