siRNA Details
| Gene Symbol: Mmp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030289.1 | NP_001025460.1 | 4, 5 | 902 | ||
| GenBank mRNA | AK155813.1 | - | 5, 6 | 889 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp27| Gene Symbol: Mmp27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030289.1 | NP_001025460.1 | 4, 5 | 902 | ||
| GenBank mRNA | AK155813.1 | - | 5, 6 | 889 | ||