siRNA Details
| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145611.4 | NP_663586.3 | 3 | 1002 | ||
| GenBank mRNA | AK031324.1 | - | 3 | 994 | ||
| AK045492.1 | - | 3 | 974 | |||
| AK049288.1 | - | 3 | 1003 | |||
| BC006647.1 | - | 3 | 990 | |||
| BC010245.1 | - | 3 | 959 | 3710765 | ||

Kank2
Genomic Map