siRNA Details
| Gene Symbol: Tmem205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253867.1 | NP_001240796.1 | 4 | 653 | ||
| NM_001253868.1 | NP_001240797.1 | 4 | 726 | |||
| NM_001253869.1 | NP_001240798.1 | 4 | 585 | |||
| NM_001253870.1 | NP_001240799.1 | 4 | 713 | |||
| NM_178577.5 | NP_848692.2 | 4 | 721 | |||
| GenBank mRNA | AB030201.1 | - | 4 | 556 | ||
| AK003909.1 | - | 1 | 745 | |||
| BC010787.1 | - | 4 | 648 | 4211629 | ||
| CA326029.1 | - | 4 | 585 | |||

Tmem205
Genomic Map