siRNA Details
| Gene Symbol: Zfp599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181419.3 | NP_852084.2 | 5 | 1643 | ||
| GenBank mRNA | AK032725.1 | - | 5 | 1594 | ||
| BC050092.1 | - | 5 | 1643 | 30019042 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp599| Gene Symbol: Zfp599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181419.3 | NP_852084.2 | 5 | 1643 | ||
| GenBank mRNA | AK032725.1 | - | 5 | 1594 | ||
| BC050092.1 | - | 5 | 1643 | 30019042 | ||