siRNA Details
| Gene Symbol: Adamts15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024139.1 | NP_001019310.1 | 7, 8 | 2302 | ||
| GenBank mRNA | BC009667.1 | - | 2, 3 | 258 | ||
| BC043308.1 | - | 5, 6 | 923 | |||
| BC094677.1 | - | 7, 8 | 2302 | 30619053 | ||

Adamts15
Genomic Map