siRNA Details
| Gene Symbol: Nfrkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172766.3 | NP_766354.2 | 23 | 3142 | ||
| GenBank mRNA | AK041206.1 | - | 23 | 3380 | ||
| AK049804.1 | - | 9 | 1517 | |||
| BC029701.1 | - | 4 | 862 | |||
| BC033595.1 | - | 23 | 3132 | 4947357 | ||
| BC059861.1 | - | 23 | 3189 | |||
| BC059900.1 | - | 14 | 2789 | |||
| BC060076.1 | - | 23 | 3371 | |||

Nfrkb
Genomic Map