siRNA Details
| Gene Symbol: Nfrkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172766.3 | NP_766354.2 | 12, 13 | 1506 | ||
| GenBank mRNA | AK041206.1 | - | 12, 13 | 1744 | ||
| BC033595.1 | - | 12, 13 | 1496 | 4947357 | ||
| BC059861.1 | - | 12, 13 | 1553 | |||
| BC059900.1 | - | 4, 5 | 418 | |||
| BC060076.1 | - | 12, 13 | 1735 | |||

Nfrkb
Genomic Map