siRNA Details
| Gene Symbol: Olfr149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207138.1 | NP_997021.1 | 1 | 806 | ||
| GenBank mRNA | BC141260.1 | - | 1 | 861 | 9055872 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr149| Gene Symbol: Olfr149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207138.1 | NP_997021.1 | 1 | 806 | ||
| GenBank mRNA | BC141260.1 | - | 1 | 861 | 9055872 | |