siRNA Details
| Gene Symbol: Gramd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172768.1 | NP_766356.1 | 15, 16 | 2128 | ||
| GenBank mRNA | AK028277.1 | - | 16, 17 | 2023 | ||
| AK033156.1 | - | 15, 16 | 2128 | |||
| AK122465.1 | - | 14, 15 | 2088 | |||
| BC030911.1 | - | 6, 7 | 872 | |||
| BC070451.1 | - | 14, 15 | 1962 | 30616509 | ||

Gramd1b
Genomic Map