siRNA Details
| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172770.3 | NP_766358.1 | 13 | 1221 | ||
| GenBank mRNA | AK054350.1 | - | 13 | 1222 | ||
| BC056616.1 | - | 12 | 1149 | 4974294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc12| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172770.3 | NP_766358.1 | 13 | 1221 | ||
| GenBank mRNA | AK054350.1 | - | 13 | 1222 | ||
| BC056616.1 | - | 12 | 1149 | 4974294 | ||