siRNA Details
| Gene Symbol: Dmxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172771.2 | NP_766359.2 | 18 | 3565 | ||
| GenBank mRNA | AK053672.1 | - | 18 | 3566 | ||
| AK170407.1 | - | 16 | 3211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmxl2| Gene Symbol: Dmxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172771.2 | NP_766359.2 | 18 | 3565 | ||
| GenBank mRNA | AK053672.1 | - | 18 | 3566 | ||
| AK170407.1 | - | 16 | 3211 | |||