siRNA Details
| Gene Symbol: Dmxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172771.2 | NP_766359.2 | 5 | 613 | ||
| GenBank mRNA | AK032099.1 | - | 5 | 559 | ||
| AK048831.1 | - | 3 | 1298 | |||
| AK053672.1 | - | 5 | 614 | |||
| AK170274.1 | - | 5 | 600 | |||
| AK170407.1 | - | 3 | 259 | |||

Dmxl2
Genomic Map