siRNA Details
| Gene Symbol: Dmxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172771.2 | NP_766359.2 | 12 | 2126 | ||
| GenBank mRNA | AK032099.1 | - | 12 | 2072 | ||
| AK053672.1 | - | 12 | 2127 | |||
| AK170274.1 | - | 12 | 2115 | |||
| AK170407.1 | - | 10 | 1772 | |||

Dmxl2
Genomic Map