siRNA Details
| Gene Symbol: Coro2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175484.3 | NP_780693.2 | 11 | 1651 | ||
| GenBank mRNA | AK047527.1 | - | 11 | 1593 | ||
| AK087414.1 | - | 11 | 1652 | |||
| BC062649.1 | - | 11 | 1383 | 6416110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coro2b| Gene Symbol: Coro2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175484.3 | NP_780693.2 | 11 | 1651 | ||
| GenBank mRNA | AK047527.1 | - | 11 | 1593 | ||
| AK087414.1 | - | 11 | 1652 | |||
| BC062649.1 | - | 11 | 1383 | 6416110 | ||