siRNA Details
| Gene Symbol: Fam63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172772.2 | NP_766360.2 | 1 | 785 | ||
| GenBank mRNA | AK035930.1 | - | 1 | 786 | ||
| AK046403.1 | - | 1 | 772 | |||
| AK046623.1 | - | 1 | 759 | |||
| AK220158.1 | - | 1 | 814 | |||
| BC049357.1 | - | 1 | 762 | |||
| BC055816.1 | - | 1 | 513 | |||
| BC058683.1 | - | 1 | 743 | 6834871 | ||
| BC069845.1 | - | 1 | 498 | |||

Fam63b
Genomic Map