siRNA Details
| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153459.4 | NP_703189.3 | 3 | 1277 | ||
| GenBank mRNA | AK140372.1 | - | 3 | 1278 | ||
| AK158817.1 | - | 2 | 453 | |||
| BC010207.1 | - | 3 | 920 | 3964527 | ||
| BC025048.1 | - | 2 | 268 | |||

Dusp7
Genomic Map