siRNA Details
| Gene Symbol: Fbxw19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177703.3 | NP_808371.1 | 9, 10 | 1310 | ||
| GenBank mRNA | AK087808.1 | - | 9, 10 | 1311 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxw19| Gene Symbol: Fbxw19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177703.3 | NP_808371.1 | 9, 10 | 1310 | ||
| GenBank mRNA | AK087808.1 | - | 9, 10 | 1311 | ||