siRNA Details
| Gene Symbol: Nbeal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183276.2 | NP_899099.2 | 36 | 6054 | ||
| GenBank mRNA | AK129158.1 | - | 12 | 2207 | ||
| BC037746.1 | - | 1 | 28 | |||
| BC042396.1 | - | 10 | 1549 | |||
| BC157956.1 | - | 36 | 6035 | 9007284 | ||
| BC172118.1 | - | 35 | 5954 | 9054404 | ||

Nbeal2
Genomic Map