siRNA Details
| Gene Symbol: Prss42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153099.1 | NP_694739.1 | 5 | 885 | ||
| GenBank mRNA | AB052292.1 | - | 5 | 885 | ||
| AK133402.1 | - | 5 | 750 | |||
| BC060984.1 | - | 5 | 904 | 6774705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss42| Gene Symbol: Prss42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153099.1 | NP_694739.1 | 5 | 885 | ||
| GenBank mRNA | AB052292.1 | - | 5 | 885 | ||
| AK133402.1 | - | 5 | 750 | |||
| BC060984.1 | - | 5 | 904 | 6774705 | ||