siRNA Details
| Gene Symbol: Prss50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146227.4 | NP_666339.2 | 1 | 463 | ||
| GenBank mRNA | AF481144.1 | - | 1 | 449 | ||
| AK045168.1 | - | 1 | 464 | |||
| BC150686.1 | - | 1 | 462 | 9087597 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss50| Gene Symbol: Prss50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146227.4 | NP_666339.2 | 1 | 463 | ||
| GenBank mRNA | AF481144.1 | - | 1 | 449 | ||
| AK045168.1 | - | 1 | 464 | |||
| BC150686.1 | - | 1 | 462 | 9087597 | ||