siRNA Details
| Gene Symbol: Rtp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153100.2 | NP_694740.2 | 3 | 1038 | ||
| GenBank mRNA | AJ428064.1 | - | 3 | 1038 | ||
| AY562227.1 | - | 3 | 936 | |||
| BC112903.1 | - | 3 | 1038 | 4196964 | ||
| BC150678.1 | - | 3 | 982 | 9087589 | ||

Rtp3
Genomic Map