siRNA Details
| Gene Symbol: Mrgpra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153101.3 | NP_694741.2 | 2 | 844 | ||
| GenBank mRNA | AY042192.1 | - | 818 | |||
| Gene Symbol: Mrgpra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172588.1 | NP_001166059.1 | 819 | |||
| GenBank mRNA | BC064040.1 | - | 819 | 1428847 | ||
More Information
Set Membership: |
|

Mrgpra2b
Genomic Map