siRNA Details
| Gene Symbol: Mrgpra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153524.2 | NP_705744.2 | 1 | 715 | ||
| GenBank mRNA | AY042194.1 | - | 1 | 715 | ||
| BC115741.1 | - | 1 | 514 | 40045594 | ||
More Information
Set Membership: |
|

Mrgpra4
Genomic Map