siRNA Details
Gene Symbol: Dhrsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033326.2 | NP_001028498.2 | 7 | 2007 | ||
GenBank mRNA | AK030649.1 | - | 6 | 1935 | ||
AK157461.1 | - | 7 | 1692 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhrsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033326.2 | NP_001028498.2 | 7 | 2007 | ||
GenBank mRNA | AK030649.1 | - | 6 | 1935 | ||
AK157461.1 | - | 7 | 1692 |