siRNA Details
| Gene Symbol: Dhrsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033326.2 | NP_001028498.2 | 7 | 2116 | ||
| GenBank mRNA | AK030649.1 | - | 6 | 2044 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrsx| Gene Symbol: Dhrsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033326.2 | NP_001028498.2 | 7 | 2116 | ||
| GenBank mRNA | AK030649.1 | - | 6 | 2044 | ||