siRNA Details
| Gene Symbol: Slc22a26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146232.1 | NP_666344.1 | 2 | 713 | ||
| GenBank mRNA | AJ132857.1 | - | 624 | |||
| BC014805.1 | - | 2 | 713 | 4242162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a26| Gene Symbol: Slc22a26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146232.1 | NP_666344.1 | 2 | 713 | ||
| GenBank mRNA | AJ132857.1 | - | 624 | |||
| BC014805.1 | - | 2 | 713 | 4242162 | ||