siRNA Details
| Gene Symbol: Tcstv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153523.3 | NP_705743.1 | 2 | 419 | ||
| GenBank mRNA | AF067060.1 | - | 417 | |||
| BC125501.1 | - | 2 | 402 | 40130016 | ||
| BC125503.1 | - | 2 | 402 | 40130018 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcstv3| Gene Symbol: Tcstv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153523.3 | NP_705743.1 | 2 | 419 | ||
| GenBank mRNA | AF067060.1 | - | 417 | |||
| BC125501.1 | - | 2 | 402 | 40130016 | ||
| BC125503.1 | - | 2 | 402 | 40130018 | ||