siRNA Details
| Gene Symbol: Lancl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173414.3 | NP_775590.2 | 3, 4 | 1159 | ||
| GenBank mRNA | AK031623.1 | - | 3, 4 | 1160 | ||
| AK043841.1 | - | 3, 4 | 360 | |||
| AK045254.1 | - | 3, 4 | 747 | |||
| BC115944.1 | - | 2, 3 | 282 | |||

Lancl3
Genomic Map