siRNA Details
| Gene Symbol: Slc22a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172776.2 | NP_766364.1 | 6, 7 | 1239 | ||
| GenBank mRNA | AF536195.1 | - | 6, 7 | 1311 | ||
| AF536196.1 | - | 7, 8 | 1047 | |||
| AK052597.1 | - | 6, 7 | 1240 | |||
| BC126952.1 | - | 7, 8 | 1052 | 40090863 | ||

Slc22a29
Genomic Map