siRNA Details
| Gene Symbol: Pyhin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175026.3 | NP_778191.1 | 6 | 1532 | ||
| GenBank mRNA | AK076726.1 | - | 2 | 410 | ||
| AK080371.1 | - | 6 | 1522 | |||
| AK150817.1 | - | 6 | 1423 | |||
| BC096384.1 | - | 6 | 1486 | 30752094 | ||
| BC107378.1 | - | 7 | 1474 | 40056703 | ||
| BC107379.1 | - | 6 | 1366 | 40056704 | ||

Pyhin1
Genomic Map