siRNA Details
| Gene Symbol: Phgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016966.3 | NP_058662.2 | 7 | 820 | ||
| GenBank mRNA | AK076815.1 | - | 7 | 776 | ||
| AK147946.1 | - | 7 | 769 | |||
| AK191824.1 | - | 2 | 109 | |||
| AK195143.1 | - | 2 | 109 | |||
| AK202836.1 | - | 2 | 109 | |||
| AK218722.1 | - | 2 | 109 | |||
| BC086668.1 | - | 7 | 728 | 30633243 | ||
| BC110673.1 | - | 7 | 762 | 6311873 | ||

Phgdh
Genomic Map