siRNA Details
| Gene Symbol: Spry3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030293.2 | NP_001025464.1 | 2 | 1261 | ||
| GenBank mRNA | AK138539.1 | - | 2 | 1261 | ||
| BC132135.1 | - | 1 | 876 | 40130412 | ||
| BC132137.1 | - | 1 | 876 | 40130414 | ||

Spry3
Genomic Map