siRNA Details
| Gene Symbol: Rbm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167775.1 | NP_001161247.1 | 22 | 3228 | ||
| NM_001167776.1 | NP_001161248.1 | 21 | 2713 | |||
| NM_145627.2 | NP_663602.1 | 22 | 2944 | |||
| GenBank mRNA | AK089105.1 | - | 21 | 2524 | ||
| AK122208.1 | - | 19 | 4644 | |||
| AK145991.1 | - | 22 | 2944 | |||
| AK153736.1 | - | 22 | 3229 | |||
| AK167666.1 | - | 21 | 2691 | |||
| BC004674.1 | - | 21 | 2629 | 3500403 | ||

Rbm10
Genomic Map