siRNA Details
| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172779.4 | NP_766367.1 | 9 | 1483 | ||
| GenBank mRNA | AK031945.1 | - | 9 | 1475 | ||
| AK083942.1 | - | 9 | 1445 | |||
| AK160627.1 | - | 9 | 1443 | |||
| BC051161.1 | - | 9 | 1465 | 6466142 | ||

Ddx26b
Genomic Map