siRNA Details
| Gene Symbol: Gm14743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126321.2 | NP_001119793.1 | 3 | 257 | ||
| GenBank mRNA | BE647313.1 | - | 2 | 59 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14743| Gene Symbol: Gm14743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126321.2 | NP_001119793.1 | 3 | 257 | ||
| GenBank mRNA | BE647313.1 | - | 2 | 59 | ||